COM945365337 Combi-Blocks, Inc.

tert-Butyl N-[(2S)-1-carbamoylpropan-2-yl]carbamate

  • MDL number MFCD30163547
  • Cas Number : 1821725-17-0
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4304

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4304
Availability details for
Quantity
  • Availability
  • Add to Cart