COM945365342 Combi-Blocks, Inc.

3-Chloro-2-fluoro-5-nitrophenylboronic acid pinacol ester

  • MDL number MFCD30726981
  • Cas Number : 2377611-03-3
  • Molecular Weight : 301.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4626
Availability details for
Quantity
  • Availability
  • Add to Cart