COM945365343 Combi-Blocks, Inc.

6-Bromo-8-fluoroquinoline-3-boronic acid pinacol ester

  • MDL number MFCD30726982
  • Cas Number : 2377608-99-4
  • Molecular Weight : 352.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5353

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5353
Availability details for
Quantity
  • Availability
  • Add to Cart