COM945365346 Combi-Blocks, Inc.

(5-Nitropyridin-2-yl)acetic acid

  • MDL number MFCD13194546
  • Cas Number : 1214328-73-0
  • Molecular Weight : 182.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7126

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7126
Availability details for
Quantity
  • Availability
  • Add to Cart