COM945365353 Combi-Blocks, Inc.

Methyl alpha-d-galactopyranoside monohydrate

  • MDL number MFCD00150748
  • Cas Number : 34004-14-3
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1842

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1842
Availability details for
Quantity
  • Availability
  • Add to Cart