COM945365371 Combi-Blocks, Inc.

(+/-)-2-Chloro-1-phenylethanol

  • MDL number MFCD01075658
  • Cas Number : 1674-30-2
  • Molecular Weight : 156.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6558

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-6558
Availability details for
Quantity
  • Availability
  • Add to Cart