COM945365384 Combi-Blocks, Inc.

Zinc carbonate hydroxide

  • MDL number MFCD00011519
  • Cas Number : 5263-02-5
  • Molecular Weight : 549.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9219

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-9219
Availability details for
Quantity
  • Availability
  • Add to Cart