COM945365416 Combi-Blocks, Inc.

3-Phenylcinnoline-4-carboxylic acid

  • MDL number MFCD00219795
  • Cas Number : 10604-21-4
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4999

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4999
Availability details for
Quantity
  • Availability
  • Add to Cart