COM945365417 Combi-Blocks, Inc.

2,2'-Bithiophene-5-methylamine

  • MDL number MFCD00110111
  • Cas Number : 4380-96-5
  • Molecular Weight : 195.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-5079

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-5079
Availability details for
Quantity
  • Availability
  • Add to Cart