COM945365419 Combi-Blocks, Inc.

1-(2-Chlorobenzyl)-1,4-diazepane

  • MDL number MFCD01312090
  • Cas Number : 76141-88-3
  • Molecular Weight : 224.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-8147

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-8147
Availability details for
Quantity
  • Availability
  • Add to Cart