COM945365426 Combi-Blocks, Inc.

[1-(6-Methylpyrazin-2-yl)piperid-3-yl]methanol

  • MDL number MFCD09879916
  • Cas Number : 937795-91-0
  • Molecular Weight : 207.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-0733
Availability details for
Quantity
  • Availability
  • Add to Cart