COM945365428 Combi-Blocks, Inc.

5-(2-Methylphenyl)-2-furoic acid

  • MDL number MFCD04039143
  • Cas Number : 80174-04-5
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2739
Availability details for
Quantity
  • Availability
  • Add to Cart