COM945365433 Combi-Blocks, Inc.

2-(4-Methylperhydro-1,4-diazepin-1-yl)benzylamine

  • MDL number MFCD09065001
  • Cas Number : 915707-56-1
  • Molecular Weight : 219.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-5132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-5132
Availability details for
Quantity
  • Availability
  • Add to Cart