COM945365441 Combi-Blocks, Inc.

4-(4-Methylperhydro-1,4-diazepin-1-yl)benzoic acid hydrochloride hydrate

  • MDL number MFCD08741428
  • Cas Number : 906352-84-9
  • Molecular Weight : 288.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8744

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-8744
Availability details for
Quantity
  • Availability
  • Add to Cart