COM945365447 Combi-Blocks, Inc.

2-bromo-1-ethyl-4-iodobenzene

  • MDL number MFCD22491026
  • Cas Number : 1369855-28-6
  • Molecular Weight : 311.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4329

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4329
Availability details for
Quantity
  • Availability
  • Add to Cart