COM945365448 Combi-Blocks, Inc.

tert-butyl N-(5-bromo-2-cyanophenyl)carbamate

  • MDL number MFCD09751792
  • Cas Number : 1260783-74-1
  • Molecular Weight : 297.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4330
Availability details for
Quantity
  • Availability
  • Add to Cart