COM945365452 Combi-Blocks, Inc.

3-Bromo-5-(pyrrolidin-1-yl)benzamide

  • MDL number MFCD30726985
  • Cas Number : 2091999-63-0
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4334

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4334
Availability details for
Quantity
  • Availability
  • Add to Cart