COM945365456 Combi-Blocks, Inc.

1-(3-Fluoro-4-piperazin-1-ylphenyl)ethanone

  • MDL number MFCD01571616
  • Cas Number : 189763-57-3
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4340
Availability details for
Quantity
  • Availability
  • Add to Cart