COM945365467 Combi-Blocks, Inc.

3-Methylnonane-2,4-dione

  • MDL number MFCD08741451
  • Cas Number : 113486-29-6
  • Molecular Weight : 170.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-9100
Availability details for
Quantity
  • Availability
  • Add to Cart