COM945365468 Combi-Blocks, Inc.

(S)-4-BOC-5-Phenylmorpholin-2-one

  • MDL number MFCD02684331
  • Cas Number : 220077-24-7
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1567

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-1567
Availability details for
Quantity
  • Availability
  • Add to Cart