COM945365469 Combi-Blocks, Inc.

3-Methoxypyrazine-2-carbonitrile

  • MDL number MFCD05863451
  • Cas Number : 75018-05-2
  • Molecular Weight : 135.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-6952
Availability details for
Quantity
  • Availability
  • Add to Cart