COM945365470 Combi-Blocks, Inc.

6-Chloroquinoline-8-carbaldehyde

  • MDL number MFCD15526831
  • Cas Number : 22934-45-8
  • Molecular Weight : 191.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2065

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2065
Availability details for
Quantity
  • Availability
  • Add to Cart