COM945365472 Combi-Blocks, Inc.

8-methoxyquinoline-4-carbonitrile

  • MDL number MFCD28337081
  • Cas Number : 1231761-22-0
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2275

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2275
Availability details for
Quantity
  • Availability
  • Add to Cart