COM945365473 Combi-Blocks, Inc.

methyl 2-methanesulfonylpyridine-4-carboxylate

  • MDL number MFCD28124339
  • Cas Number : 1637781-26-0
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2292
Availability details for
Quantity
  • Availability
  • Add to Cart