COM945365474 Combi-Blocks, Inc.

5-bromo-4-chloropyrimidin-2-ol

  • MDL number MFCD16988221
  • Cas Number : 1240595-19-0
  • Molecular Weight : 209.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2305

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2305
Availability details for
Quantity
  • Availability
  • Add to Cart