COM945365475 Combi-Blocks, Inc.

1H-Pyrazolo[3,4-d]pyrimidin-3-amine

  • MDL number MFCD14713511
  • Cas Number : 875215-47-7
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2309

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2309
Availability details for
Quantity
  • Availability
  • Add to Cart