COM945365476 Combi-Blocks, Inc.

3-Iodo-1H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD11040174
  • Cas Number : 144750-83-4
  • Molecular Weight : 262.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2317

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2317
Availability details for
Quantity
  • Availability
  • Add to Cart