COM945365477 Combi-Blocks, Inc.

Imidazo[1,2-a]pyridine-2-carboxamide

  • MDL number MFCD11643701
  • Cas Number : 39031-44-2
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2321

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2321
Availability details for
Quantity
  • Availability
  • Add to Cart