COM945365479 Combi-Blocks, Inc.

4,6-Bis(methylsulfanyl)-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD00619052
  • Cas Number : 6288-89-7
  • Molecular Weight : 212.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2326
Availability details for
Quantity
  • Availability
  • Add to Cart