COM945365481 Combi-Blocks, Inc.

4-(Dimethoxymethyl)benzoic acid

  • MDL number MFCD11554157
  • Cas Number : 120465-56-7
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6327
Availability details for
Quantity
  • Availability
  • Add to Cart