COM945365484 Combi-Blocks, Inc.

3-Chloro-4-(hydroxymethyl)benzoic acid

  • MDL number MFCD22491677
  • Cas Number : 1594779-14-2
  • Molecular Weight : 186.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6365

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6365
Availability details for
Quantity
  • Availability
  • Add to Cart