COM945365491 Combi-Blocks, Inc.

4-Chloro-5-(trimethylsilyl)thiophene-2-carbaldehyde

  • MDL number MFCD30726991
  • Cas Number : 2092000-97-8
  • Molecular Weight : 218.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6425
Availability details for
Quantity
  • Availability
  • Add to Cart