COM945365496 Combi-Blocks, Inc.

1-(4-Cyanophenyl)methanesulfonamide

  • MDL number MFCD07689448
  • Cas Number : 191868-54-9
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3951

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3951
Availability details for
Quantity
  • Availability
  • Add to Cart