COM945365497 Combi-Blocks, Inc.

(4-Aminomethylbenzyl)diethylamine

  • MDL number MFCD07343995
  • Cas Number : 84227-70-3
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6479

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-6479
Availability details for
Quantity
  • Availability
  • Add to Cart