COM945365498 Combi-Blocks, Inc.

1-(2-Cyanophenyl)methanesulfonamide

  • MDL number MFCD07689449
  • Cas Number : 27350-13-6
  • Molecular Weight : 196.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9970

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-9970
Availability details for
Quantity
  • Availability
  • Add to Cart