COM945365500 Combi-Blocks, Inc.

Methyl 2-(3-cyanophenyl)benzoate

  • MDL number MFCD05978798
  • Cas Number : 352616-88-7
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4482

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4482
Availability details for
Quantity
  • Availability
  • Add to Cart