COM945512093 Combi-Blocks, Inc.

2-(4-Methyl-1,4-diazepan-1-yl)benzonitrile

  • MDL number MFCD09065002
  • Cas Number : 204078-93-3
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2496
Availability details for
Quantity
  • Availability
  • Add to Cart