COM945512129 Combi-Blocks, Inc.

3,5-Dibromo-2,6-difluorobenzoic acid

  • MDL number MFCD28783383
  • Cas Number : 28314-84-3
  • Molecular Weight : 315.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5959
Availability details for
Quantity
  • Availability
  • Add to Cart