COM945512133 Combi-Blocks, Inc.

(3-Aminomethylphenyl)boronic acid

  • MDL number MFCD06212999
  • Cas Number : 146285-80-5
  • Molecular Weight : 151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2627

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2627
Availability details for
Quantity
  • Availability
  • Add to Cart