COM945512149 Combi-Blocks, Inc.

(4-methyl-1,3-thiazol-2-yl)methanamine HCl

  • MDL number MFCD10697495
  • Cas Number : 1196146-15-2
  • Molecular Weight : 164.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4076
Availability details for
Quantity
  • Availability
  • Add to Cart