COM945512173 Combi-Blocks, Inc.

1-(2-Cyanophenyl)-N-(2-methoxyethyl)methanesulfonamide

  • MDL number MFCD11123401
  • Cas Number : 1041527-14-3
  • Molecular Weight : 254.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1745
Availability details for
Quantity
  • Availability
  • Add to Cart