COM945512201 Combi-Blocks, Inc.

1-[4-(aminomethyl)phenyl]-N-phenylmethanesulfonamide

  • MDL number MFCD12443452
  • Cas Number : 1041556-68-6
  • Molecular Weight : 276.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1797
Availability details for
Quantity
  • Availability
  • Add to Cart