COM945512206 Combi-Blocks, Inc.

2-(4-Cyano-2-nitrophenyl)acetamide

  • MDL number MFCD30834378
  • Cas Number : 2140305-61-7
  • Molecular Weight : 205.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1803

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1803
Availability details for
Quantity
  • Availability
  • Add to Cart