COM945512216 Combi-Blocks, Inc.

7-tert-butyl-1H-pyrazolo[1,5-a][1,3,5]triazin-2-one

  • MDL number MFCD30834385
  • Cas Number : 2140305-49-1
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1814

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1814
Availability details for
Quantity
  • Availability
  • Add to Cart