COM945512224 Combi-Blocks, Inc.

5-methyl-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD07440139
  • Cas Number : 27582-24-7
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1864
Availability details for
Quantity
  • Availability
  • Add to Cart