COM945512265 Combi-Blocks, Inc.

tert-Butyl 4-bromo-6-hydroxy-1-oxo-3H-isoindole-2-carboxylate

  • MDL number MFCD30527655
  • Cas Number : 2055119-08-7
  • Molecular Weight : 328.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4230

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4230
Availability details for
Quantity
  • Availability
  • Add to Cart