COM945512287 Combi-Blocks, Inc.

1-methanesulfonyl-4-phenylpiperidine

  • MDL number MFCD00957567
  • Cas Number : 59039-19-9
  • Molecular Weight : 239.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4337
Availability details for
Quantity
  • Availability
  • Add to Cart