COM945512331 Combi-Blocks, Inc.

2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid hydrate

  • MDL number MFCD00270531
  • Cas Number : 18835-59-1
  • Molecular Weight : 451

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3640

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3640
Availability details for
Quantity
  • Availability
  • Add to Cart