COM945512337 Combi-Blocks, Inc.

4-Amino-6-hydroxy-2-methylpyrimidine dihydrate

  • MDL number MFCD00150311
  • Cas Number : 388582-41-0
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4466
Availability details for
Quantity
  • Availability
  • Add to Cart