COM945512504 Combi-Blocks, Inc.

4-(Thien-2-yl)aniline hydrochloride

  • MDL number MFCD00136945
  • Cas Number : 1172363-16-4
  • Molecular Weight : 211.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4881

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-4881
Availability details for
Quantity
  • Availability
  • Add to Cart